Product Details

SNP ID
rs117602546
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:116673656 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACATTATATTCCCTTACAGCTAAA[C/T]TATTATAGCTGACTAATGAATTTTT
Phenotype
MIM: 164860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MET PubMed Links

Gene Details

Gene
MET
Gene Name
MET proto-oncogene, receptor tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000245.3 Intron NP_000236.2
NM_001127500.2 Intron NP_001120972.1
NM_001324401.1 Intron NP_001311330.1
NM_001324402.1 Intron NP_001311331.1
XM_006715990.2 Intron XP_006716053.1
XM_011516223.1 Intron XP_011514525.1

View Full Product Details