Product Details

SNP ID
rs117483380
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:121328620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGAGGTGAAGGAGAAGAAAAAAA[A/G]GTCAGTGACATTTTCCAGCCACTAC
Phenotype
MIM: 606267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
WNT16 PubMed Links

Gene Details

Gene
WNT16
Gene Name
Wnt family member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016087.2 Intron NP_057171.2
NM_057168.1 Intron NP_476509.1

View Full Product Details