Product Details

SNP ID
rs112299567
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:6533061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTTCAGAAAATGGAGACTCATAAA[C/T]TTCCATGGGTGGGCAGGTACAAACC
Phenotype
MIM: 238300
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GLDC PubMed Links

Gene Details

Gene
GLDC
Gene Name
glycine decarboxylase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000170.2 Intron NP_000161.2

View Full Product Details