Product Details

SNP ID
rs113652009
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:567196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCAGGGATCCCACCTGGTGTAAGTA[C/T]GGGCAGAGGGCAGAGCCAGGCTGTA
Phenotype
MIM: 612982 MIM: 611780 MIM: 143023
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LMNTD2 PubMed Links

Gene Details

Gene
LMNTD2
Gene Name
lamin tail domain containing 2
There are no transcripts associated with this gene.

Gene
LOC143666
Gene Name
uncharacterized LOC143666
There are no transcripts associated with this gene.

Gene
MIR210
Gene Name
microRNA 210
There are no transcripts associated with this gene.

Gene
MIR210HG
Gene Name
MIR210 host gene
There are no transcripts associated with this gene.

Gene
PHRF1
Gene Name
PHD and ring finger domains 1
There are no transcripts associated with this gene.

Gene
RASSF7
Gene Name
Ras association domain family member 7
There are no transcripts associated with this gene.

View Full Product Details