Product Details
- SNP ID
-
rs113595629
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.11:6431040 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAAGAAGATTGTTTCACGACCAAG[G/T]TCCCTTGATTCAAGTGAAGAAGCAA
- Phenotype
-
MIM: 142290
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
HPX
PubMed Links
Gene Details
- Gene
- HPX
- Gene Name
- hemopexin
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000613.2 |
1621 |
UTR 3 |
|
|
NP_000604.1 |
View Full Product Details