Product Details

SNP ID
rs115662875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:122777757 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACATTGATGGGCTCATAGGGAAGAT[A/G]GAGAGGAAGTAAAGACTGCAATCCC
Phenotype
MIM: 613481 MIM: 604550
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC62 PubMed Links

Gene Details

Gene
CCDC62
Gene Name
coiled-coil domain containing 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201435.4 Intron NP_958843.2
XM_006719643.1 Intron XP_006719706.1
XM_006719644.1 Intron XP_006719707.1
XM_011538856.1 Intron XP_011537158.1
XM_011538857.1 Intron XP_011537159.1
XM_011538859.1 Intron XP_011537161.1
XM_017020033.1 Intron XP_016875522.1
Gene
DENR
Gene Name
density regulated re-initiation and release factor
There are no transcripts associated with this gene.

View Full Product Details