Product Details

SNP ID
rs117769740
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:190541 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACTCGGAGGAAACTCCAGACTGA[C/T]GGCATGCAAAGGCTTGCCTCTGTCG
Phenotype
MIM: 603080
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLC6A12 PubMed Links

Gene Details

Gene
SLC6A12
Gene Name
solute carrier family 6 member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001122847.2 3132 UTR 3 NP_001116319.1
NM_001122848.2 3132 UTR 3 NP_001116320.1
NM_001206931.1 3132 UTR 3 NP_001193860.1
NM_003044.4 3132 UTR 3 NP_003035.3
XM_005253747.4 3132 Intron XP_005253804.1
XM_005253748.4 3132 UTR 3 XP_005253805.1
XM_006719005.3 3132 Intron XP_006719068.1
XM_011521010.1 3132 UTR 3 XP_011519312.1
XM_017019841.1 3132 UTR 3 XP_016875330.1

View Full Product Details