Product Details

SNP ID
rs117715420
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2584878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTATGCAGAGGCCTGGGAGCCTC[A/G]AGGCCCTTGCCCCCACCCTGTGTGC
Phenotype
MIM: 605213
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
PDPK1 PubMed Links

Gene Details

Gene
PDPK1
Gene Name
3-phosphoinositide dependent protein kinase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001261816.1 Intron NP_001248745.1
NM_002613.4 Intron NP_002604.1
NM_031268.5 Intron NP_112558.2
XM_011522521.2 Intron XP_011520823.1
XM_011522523.2 Intron XP_011520825.1
XM_017023271.1 Intron XP_016878760.1
XM_017023272.1 Intron XP_016878761.1
XM_017023273.1 Intron XP_016878762.1
XM_017023274.1 Intron XP_016878763.1
XM_017023275.1 Intron XP_016878764.1
XM_017023276.1 Intron XP_016878765.1
XM_017023277.1 Intron XP_016878766.1

View Full Product Details