Product Details

SNP ID
rs114794799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3013422 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGGCTGTGCTGGGCTGGCTGGG[C/G]ACCCTGGTGTCCTGCGCCCTGCCCC
Phenotype
MIM: 615798 MIM: 615799 MIM: 605914
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CLDN6 PubMed Links

Gene Details

Gene
CLDN6
Gene Name
claudin 6
There are no transcripts associated with this gene.

Gene
CLDN9
Gene Name
claudin 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020982.3 967 Silent Mutation GGC,GGG G20G NP_066192.1
Gene
HCFC1R1
Gene Name
host cell factor C1 regulator 1
There are no transcripts associated with this gene.

Gene
TNFRSF12A
Gene Name
TNF receptor superfamily member 12A
There are no transcripts associated with this gene.

View Full Product Details