Product Details

SNP ID
rs116545155
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3022800 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGAGGAACCTTGTCCCTTTGCGG[C/G]AGTCGCTGGTCTCTTCTGTTGTGGG
Phenotype
MIM: 617003 MIM: 615798 MIM: 615799 MIM: 615403 MIM: 605914
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BICDL2 PubMed Links

Gene Details

Gene
BICDL2
Gene Name
BICD family like cargo adaptor 2
There are no transcripts associated with this gene.

Gene
CLDN6
Gene Name
claudin 6
There are no transcripts associated with this gene.

Gene
CLDN9
Gene Name
claudin 9
There are no transcripts associated with this gene.

Gene
HCFC1R1
Gene Name
host cell factor C1 regulator 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002017.2 392 UTR 3 NP_001002017.1
NM_001002018.2 392 UTR 3 NP_001002018.1
NM_001288665.1 392 UTR 3 NP_001275594.1
NM_001288666.1 392 UTR 3 NP_001275595.1
NM_001288667.1 392 UTR 3 NP_001275596.1
NM_001288668.1 392 UTR 3 NP_001275597.1
NM_001308070.1 392 UTR 3 NP_001294999.1
NM_017885.3 392 UTR 3 NP_060355.1
XM_011522559.2 392 UTR 3 XP_011520861.1
XM_017023384.1 392 UTR 3 XP_016878873.1
Gene
LOC100128770
Gene Name
uncharacterized LOC100128770
There are no transcripts associated with this gene.

Gene
THOC6
Gene Name
THO complex 6
There are no transcripts associated with this gene.

Gene
TNFRSF12A
Gene Name
TNF receptor superfamily member 12A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016639.2 392 Intron NP_057723.1

View Full Product Details