Product Details

SNP ID
rs112538377
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:4609950 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGTGAGAGCGTCAGGGAAGTTGG[A/G]GGGTGTAGCAGGGGTATTGGCGGGG
Phenotype
MIM: 607559
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C16orf96 PubMed Links

Gene Details

Gene
C16orf96
Gene Name
chromosome 16 open reading frame 96
There are no transcripts associated with this gene.

Gene
MGRN1
Gene Name
mahogunin ring finger 1
There are no transcripts associated with this gene.

Gene
UBALD1
Gene Name
UBA like domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145253.2 346 Missense Mutation CCC,TCC P73S NP_660296.1
XM_017022937.1 346 Intron XP_016878426.1

View Full Product Details