Product Details

SNP ID
rs116147293
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:3433517 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCACAAGGAGGAAGCTCTCTAGAT[A/C]CGAAAAATACAAGAAGAAGTACATT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
OR1E2 PubMed Links

Gene Details

Gene
OR1E2
Gene Name
olfactory receptor family 1 subfamily E member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003554.2 325 Missense Mutation GAT,TAT D109Y NP_003545.1
Gene
SPATA22
Gene Name
spermatogenesis associated 22
There are no transcripts associated with this gene.

View Full Product Details