Product Details

SNP ID
rs111722981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:39904882 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGACAACAACATACAGCGCACAG[A/G]GAATTCGTGCCTCAGGGTCATAGTC
Phenotype
MIM: 611221
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GSDMB PubMed Links

Gene Details

Gene
GSDMB
Gene Name
gasdermin B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042471.1 3203 Missense Mutation CCC,CTC P381L NP_001035936.1
NM_001165958.1 3203 Missense Mutation CCC,CTC P394L NP_001159430.1
NM_001165959.1 3203 Missense Mutation CCC,CTC P385L NP_001159431.1
NM_018530.2 3203 Missense Mutation CCC,CTC P372L NP_061000.2
XM_011525004.1 3203 Missense Mutation CCC,CTC P398L XP_011523306.1
XM_011525006.1 3203 Missense Mutation CCC,CTC P398L XP_011523308.1
XM_011525007.2 3203 Missense Mutation CCC,CTC P398L XP_011523309.1
XM_011525009.1 3203 Missense Mutation CCC,CTC P398L XP_011523311.1
XM_011525011.1 3203 Missense Mutation CCC,CTC P398L XP_011523313.1
XM_011525012.1 3203 Missense Mutation CCC,CTC P398L XP_011523314.1
XM_011525013.1 3203 Missense Mutation CCC,CTC P398L XP_011523315.1
XM_011525015.1 3203 Missense Mutation CCC,CTC P398L XP_011523317.1
XM_017024848.1 3203 Missense Mutation CCC,CTC P398L XP_016880337.1
XM_017024849.1 3203 Missense Mutation CCC,CTC P398L XP_016880338.1
XM_017024850.1 3203 Missense Mutation CCC,CTC P394L XP_016880339.1
XM_017024851.1 3203 Missense Mutation CCC,CTC P389L XP_016880340.1
XM_017024852.1 3203 Missense Mutation CCC,CTC P381L XP_016880341.1
XM_017024853.1 3203 Intron XP_016880342.1
XM_017024854.1 3203 Intron XP_016880343.1

View Full Product Details