Product Details
- SNP ID
-
rs113361561
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:75012705 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGAGCATGGCGGCCACCAGGTGC[C/T]TGCGCTGGGGCCTGAGCCGAGCCGG
- Phenotype
-
MIM: 603000
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CDR2L
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs3744206] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CDR2L
- Gene Name
- cerebellar degeneration related protein 2 like
There are no transcripts associated with this gene.
- Gene
- MRPL58
- Gene Name
- mitochondrial ribosomal protein L58
View Full Product Details