Product Details

SNP ID
rs117785251
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75591675 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGATCATGCAGTTCCTAAGCA[G/T]CCTGGAGCAGGATCAGACGGGGAAC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MYO15B PubMed Links

Gene Details

Gene
MYO15B
Gene Name
myosin XVB
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001309242.1 2369 Missense Mutation AGC,ATC S837I NP_001296171.1
XM_017025120.1 2369 Missense Mutation AGC,ATC S619I XP_016880609.1
XM_017025121.1 2369 Missense Mutation AGC,ATC S614I XP_016880610.1
XM_017025122.1 2369 Missense Mutation AGC,ATC S619I XP_016880611.1
XM_017025123.1 2369 Missense Mutation AGC,ATC S619I XP_016880612.1
XM_017025124.1 2369 Missense Mutation AGC,ATC S619I XP_016880613.1
XM_017025125.1 2369 Missense Mutation AGC,ATC S619I XP_016880614.1
XM_017025126.1 2369 Missense Mutation AGC,ATC S619I XP_016880615.1
XM_017025127.1 2369 Missense Mutation AGC,ATC S390I XP_016880616.1
XM_017025128.1 2369 Missense Mutation AGC,ATC S381I XP_016880617.1
XM_017025129.1 2369 Missense Mutation AGC,ATC S98I XP_016880618.1
XM_017025130.1 2369 Missense Mutation AGC,ATC S90I XP_016880619.1
XM_017025131.1 2369 Missense Mutation AGC,ATC S55I XP_016880620.1
XM_017025132.1 2369 UTR 5 XP_016880621.1
XM_017025133.1 2369 UTR 5 XP_016880622.1
XM_017025134.1 2369 UTR 5 XP_016880623.1
XM_017025135.1 2369 Intron XP_016880624.1
XM_017025136.1 2369 Intron XP_016880625.1
XM_017025137.1 2369 Intron XP_016880626.1
XM_017025138.1 2369 Intron XP_016880627.1
XM_017025139.1 2369 Intron XP_016880628.1
XM_017025140.1 2369 Intron XP_016880629.1
XM_017025141.1 2369 Missense Mutation AGC,ATC S619I XP_016880630.1
XM_017025142.1 2369 Missense Mutation AGC,ATC S619I XP_016880631.1
XM_017025143.1 2369 Intron XP_016880632.1
XM_017025144.1 2369 UTR 5 XP_016880633.1

View Full Product Details