Product Details
- SNP ID
-
rs112120285
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:40960326 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGGCTGCGGTATGTGGTAATCTCA[C/G]ATTCCAGCCGGGACTTGACGTCCAG
- Phenotype
-
MIM: 616678
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
KRT39
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs17843023] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- KRT39
- Gene Name
- keratin 39
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_213656.3 |
1208 |
Missense Mutation |
TCT,TGT |
S391C |
NP_998821.3 |
View Full Product Details