Product Details

SNP ID
rs117519127
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:81228351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGTTTTTCTGAGATGGACCA[C/G]AGGCTCCAATCAGAGGCTTCGTGTG
Phenotype
MIM: 613479
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CEP131 PubMed Links

Gene Details

Gene
CEP131
Gene Name
centrosomal protein 131
There are no transcripts associated with this gene.

Gene
LOC105371925
Gene Name
uncharacterized LOC105371925
There are no transcripts associated with this gene.

Gene
TEPSIN
Gene Name
TEPSIN, adaptor related protein complex 4 accessory protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144679.2 3458 UTR 3 NP_653280.1
XM_005257066.2 3458 UTR 3 XP_005257123.1
XM_005257067.2 3458 UTR 3 XP_005257124.1
XM_006721709.3 3458 UTR 3 XP_006721772.1
XM_006721712.2 3458 UTR 3 XP_006721775.1
XM_011524355.1 3458 UTR 3 XP_011522657.1
XM_011524356.1 3458 UTR 3 XP_011522658.1
XM_011524357.1 3458 UTR 3 XP_011522659.1
XM_011524358.2 3458 UTR 3 XP_011522660.1
XM_017024202.1 3458 UTR 3 XP_016879691.1
XM_017024203.1 3458 UTR 3 XP_016879692.1
XM_017024204.1 3458 UTR 3 XP_016879693.1

View Full Product Details