Product Details
- SNP ID
-
rs112384377
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:42761319 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCGCGTCTCCCTAGGACCCGAGTC[C/G]GGCGGCCGGCAGCGCTCCGCCTCCT
- Phenotype
-
MIM: 605154
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RAMP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs753152] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RAMP2
- Gene Name
- receptor activity modifying protein 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005854.2 |
126 |
Missense Mutation |
CGG,GGG |
R20G |
NP_005845.2 |
- Gene
- RAMP2-AS1
- Gene Name
- RAMP2 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details