Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_022462.4 | 35 | Intron | NP_071907.4 | ||
NM_152794.3 | 35 | Intron | NP_690007.1 | ||
NM_152795.3 | 35 | Missense Mutation | CAG,CTG | Q3L | NP_690008.2 |
NM_152796.2 | 35 | Intron | NP_690009.1 | ||
XM_005259152.4 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_005259209.1 |
XM_005259153.3 | 35 | Intron | XP_005259210.2 | ||
XM_005259155.4 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_005259212.1 |
XM_005259156.4 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_005259213.1 |
XM_017027132.1 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_016882621.1 |
XM_017027133.1 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_016882622.1 |
XM_017027134.1 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_016882623.1 |
XM_017027135.1 | 35 | Intron | XP_016882624.1 | ||
XM_017027136.1 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_016882625.1 |
XM_017027137.1 | 35 | Intron | XP_016882626.1 | ||
XM_017027138.1 | 35 | Missense Mutation | CAG,CTG | Q3L | XP_016882627.1 |
XM_017027139.1 | 35 | Intron | XP_016882628.1 | ||
XM_017027140.1 | 35 | UTR 5 | XP_016882629.1 | ||
XM_017027141.1 | 35 | Intron | XP_016882630.1 | ||
XM_017027142.1 | 35 | Intron | XP_016882631.1 |