Product Details

SNP ID
rs115473175
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:57507635 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTATTCAACACCAGAAAGTTTAGA[A/C]TGGAGAAAGGCCTTAGACTGTCGCT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ZNF773 PubMed Links

Gene Details

Gene
ZNF773
Gene Name
zinc finger protein 773
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304334.1 1764 UTR 3 NP_001291263.1
NM_001304335.1 1764 Intron NP_001291264.1
NM_001304336.1 1764 UTR 3 NP_001291265.1
NM_001304337.1 1764 Intron NP_001291266.1
NM_198542.2 1764 UTR 3 NP_940944.1

View Full Product Details