Product Details

SNP ID
rs118158038
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:33378894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTTTTGTTTTCCTCCCATTTTTG[A/G]TAACAATCGCAGAGGGGCAGTGCCC
Phenotype
MIM: 138972 MIM: 613230
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEBPG PubMed Links

Gene Details

Gene
CEBPG
Gene Name
CCAAT/enhancer binding protein gamma
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252296.1 Intron NP_001239225.1
NM_001806.3 Intron NP_001797.1
Gene
PEPD
Gene Name
peptidase D
There are no transcripts associated with this gene.

View Full Product Details