Product Details

SNP ID
rs112254603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37150343 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAATCAATAAGGAGGATCACAAGA[C/T]ATCCTGCTAGAGGTGTAATAAAAGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF585A PubMed Links

Gene Details

Gene
ZNF585A
Gene Name
zinc finger protein 585A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001288800.1 3815 UTR 3 NP_001275729.1
NM_152655.3 3815 UTR 3 NP_689868.1
NM_199126.2 3815 UTR 3 NP_954577.1

View Full Product Details