Product Details

SNP ID
rs113145524
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:37181877 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGTAGGCGAAACACAGAGGATTA[C/T]TGGGACAGTGAAACGGCTCTTTATC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ZNF585A PubMed Links

Gene Details

Gene
ZNF585A
Gene Name
zinc finger protein 585A
There are no transcripts associated with this gene.

Gene
ZNF585B
Gene Name
zinc finger protein 585B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152279.3 5912 UTR 3 NP_689492.3

View Full Product Details