Product Details

SNP ID
rs114881279
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:21292969 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCATTTTTAAAGTTCCTCACCAGT[A/G]TGATTTATTTTATGTTTAGAAAAGT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZNF708 PubMed Links

Gene Details

Gene
ZNF708
Gene Name
zinc finger protein 708
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001297560.1 1761 Intron NP_001284489.1
NM_001297561.1 1761 Intron NP_001284490.1
NM_021269.2 1761 Intron NP_067092.2
XM_017027201.1 1761 Intron XP_016882690.1
XM_017027202.1 1761 UTR 3 XP_016882691.1
XM_017027203.1 1761 UTR 3 XP_016882692.1
XM_017027204.1 1761 Silent Mutation CAC,CAT H582H XP_016882693.1

View Full Product Details