Product Details

SNP ID
rs112370660
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63910564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCGAAAAAAAAAAGTGATGTGCG[C/T]TTGGGACTGGGAGGGCGAGGCCCGA
Phenotype
MIM: 611203
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJC5 PubMed Links

Gene Details

Gene
DNAJC5
Gene Name
DnaJ heat shock protein family (Hsp40) member C5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025219.2 Intron NP_079495.1
XM_011529048.2 Intron XP_011527350.1
XM_011529049.1 Intron XP_011527351.1
Gene
MIR941-1
Gene Name
microRNA 941-1
There are no transcripts associated with this gene.

Gene
MIR941-2
Gene Name
microRNA 941-2
There are no transcripts associated with this gene.

Gene
MIR941-3
Gene Name
microRNA 941-3
There are no transcripts associated with this gene.

Gene
MIR941-4
Gene Name
microRNA 941-4
There are no transcripts associated with this gene.

Gene
MIR941-5
Gene Name
microRNA 941-5
There are no transcripts associated with this gene.

View Full Product Details