Product Details

SNP ID
rs115571516
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.20:63006313 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGCAGCAGCGTGGCGATCTTGGA[C/G]AGCTTGCGCACCGACGGGCTGTGCG
Phenotype
MIM: 609331
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
BHLHE23 PubMed Links

Gene Details

Gene
BHLHE23
Gene Name
basic helix-loop-helix family member e23
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_080606.3 723 Silent Mutation CTC,CTG L154L NP_542173.2
Gene
LOC63930
Gene Name
uncharacterized LOC63930
There are no transcripts associated with this gene.

View Full Product Details