Product Details

SNP ID
rs115458101
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.21:33432763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGATCTCCGTGGGAACATTTTC[A/G]TTGCTGTCGGTGCTGGCAGGAGCCT
Phenotype
MIM: 147569
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IFNGR2 PubMed Links

Gene Details

Gene
IFNGR2
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005534.3 1419 Silent Mutation TCA,TCG S257S NP_005525.2
XM_005260969.2 1419 Silent Mutation TCA,TCG S276S XP_005261026.1
XM_011529553.1 1419 Silent Mutation TCA,TCG S282S XP_011527855.1
XM_011529554.2 1419 Silent Mutation TCA,TCG S259S XP_011527856.1
Gene
TMEM50B
Gene Name
transmembrane protein 50B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006134.6 1419 Intron NP_006125.2
XM_011529746.1 1419 UTR 3 XP_011528048.1

View Full Product Details