Product Details
- SNP ID
-
rs112983173
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.22:21958634 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTCCTTGTAGAGCTTGATGGTGCC[A/G]TTCTGGGGGAGCGTGTAGGTCTCAT
- Phenotype
-
MIM: 603582
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PPM1F
PubMed Links
Gene Details
- Gene
- PPM1F
- Gene Name
- protein phosphatase, Mg2+/Mn2+ dependent 1F
There are no transcripts associated with this gene.
- Gene
- TOP3B
- Gene Name
- topoisomerase (DNA) III beta
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001282112.1 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
NP_001269041.1 |
NM_001282113.1 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
NP_001269042.1 |
NM_003935.4 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
NP_003926.1 |
XM_005261811.1 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
XP_005261868.1 |
XM_005261813.2 |
2218 |
Silent Mutation |
AAC,AAT |
N519N |
XP_005261870.1 |
XM_006724349.1 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
XP_006724412.1 |
XM_006724350.1 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
XP_006724413.1 |
XM_011530482.1 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
XP_011528784.1 |
XM_017029038.1 |
2218 |
Silent Mutation |
AAC,AAT |
N655N |
XP_016884527.1 |
XM_017029039.1 |
2218 |
Silent Mutation |
AAC,AAT |
N519N |
XP_016884528.1 |
XM_017029040.1 |
2218 |
Silent Mutation |
AAC,AAT |
N519N |
XP_016884529.1 |
XM_017029041.1 |
2218 |
Silent Mutation |
AAC,AAT |
N519N |
XP_016884530.1 |
View Full Product Details