Product Details

SNP ID
rs115172371
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:42012646 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTATTTGGTAATGTCTTAATTTCT[C/T]TTTAATTTTTGAAGGATAGTTTTGC
Phenotype
MIM: 610981
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WBP2NL PubMed Links

Gene Details

Gene
WBP2NL
Gene Name
WBP2 N-terminal like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152613.2 Intron NP_689826.2

View Full Product Details