Product Details

SNP ID
rs121434615
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.X:120539349 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTAGCTTCTTTGTTGCCTGCAC[G/A]AAGTTTTGAATCTACATACTTAGCT
Phenotype
MIM: 300304
Polymorphism
G/A, Transition Substitution
Allele Nomenclature
Literature Links
CUL4B PubMed Links

Gene Details

Gene
CUL4B
Gene Name
cullin 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001079872.1 1984 Missense Mutation CGT,TGT R554C NP_001073341.1
NM_003588.3 1984 Missense Mutation CGT,TGT R572C NP_003579.3
XM_005262481.1 1984 Missense Mutation CGT,TGT R572C XP_005262538.1
XM_006724784.1 1984 Missense Mutation CGT,TGT R559C XP_006724847.1
XM_006724785.2 1984 Missense Mutation CGT,TGT R559C XP_006724848.1
XM_011531399.2 1984 Missense Mutation CGT,TGT R376C XP_011529701.1
XM_011531400.2 1984 Missense Mutation CGT,TGT R376C XP_011529702.1
XM_011531401.1 1984 Missense Mutation CGT,TGT R358C XP_011529703.1

View Full Product Details