Product Details

SNP ID
rs113407270
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:177431726 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGATGGATTAGATGCAGTCCACTTT[C/T]TCCTCAGTTGGCTGATTTTTTTCGG
Phenotype
MIM: 613228
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AGA PubMed Links

Gene Details

Gene
AGA
Gene Name
aspartylglucosaminidase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000027.3 1121 Silent Mutation GAA,GAG E341E NP_000018.2
NM_001171988.1 1121 Silent Mutation GAA,GAG E331E NP_001165459.1

View Full Product Details