Product Details

SNP ID
rs116639941
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:112724380 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCCGGCCAGAAAAATCTTGAAA[A/G]TGTGAGATTATTGATTCATTTGACA
Phenotype
MIM: 106410
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ANK2 PubMed Links

Gene Details

Gene
ANK2
Gene Name
ankyrin 2, neuronal
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127493.1 Intron NP_001120965.1
NM_001148.4 Intron NP_001139.3
NM_020977.3 Intron NP_066187.2
XM_005262945.2 Intron XP_005263002.1
XM_017008067.1 Intron XP_016863556.1
XM_017008068.1 Intron XP_016863557.1
XM_017008069.1 Intron XP_016863558.1
XM_017008070.1 Intron XP_016863559.1
XM_017008071.1 Intron XP_016863560.1
XM_017008072.1 Intron XP_016863561.1
XM_017008073.1 Intron XP_016863562.1
XM_017008074.1 Intron XP_016863563.1
XM_017008075.1 Intron XP_016863564.1
XM_017008076.1 Intron XP_016863565.1
XM_017008077.1 Intron XP_016863566.1
XM_017008078.1 Intron XP_016863567.1
XM_017008079.1 Intron XP_016863568.1
XM_017008080.1 Intron XP_016863569.1
XM_017008081.1 Intron XP_016863570.1
XM_017008082.1 Intron XP_016863571.1
XM_017008083.1 Intron XP_016863572.1
XM_017008084.1 Intron XP_016863573.1
XM_017008085.1 Intron XP_016863574.1
XM_017008086.1 Intron XP_016863575.1
XM_017008087.1 Intron XP_016863576.1
XM_017008088.1 Intron XP_016863577.1
XM_017008089.1 Intron XP_016863578.1
XM_017008090.1 Intron XP_016863579.1
XM_017008091.1 Intron XP_016863580.1
XM_017008092.1 Intron XP_016863581.1
XM_017008093.1 Intron XP_016863582.1
XM_017008094.1 Intron XP_016863583.1
XM_017008095.1 Intron XP_016863584.1
XM_017008096.1 Intron XP_016863585.1
XM_017008097.1 Intron XP_016863586.1
XM_017008098.1 Intron XP_016863587.1
XM_017008099.1 Intron XP_016863588.1
XM_017008100.1 Intron XP_016863589.1
XM_017008101.1 Intron XP_016863590.1
XM_017008102.1 Intron XP_016863591.1
XM_017008103.1 Intron XP_016863592.1
XM_017008104.1 Intron XP_016863593.1
XM_017008105.1 Intron XP_016863594.1
XM_017008106.1 Intron XP_016863595.1
XM_017008107.1 Intron XP_016863596.1
XM_017008108.1 Intron XP_016863597.1
XM_017008109.1 Intron XP_016863598.1
XM_017008110.1 Intron XP_016863599.1
XM_017008111.1 Intron XP_016863600.1
XM_017008112.1 Intron XP_016863601.1

View Full Product Details