Product Details

SNP ID
rs117703219
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49198436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCCAACTCCTTGGCTGCTTTTCC[C/T]TCATCTCCAAAGTCCTCTAAACTGG
Phenotype
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
C3orf84 PubMed Links

Gene Details

Gene
C3orf84
Gene Name
chromosome 3 open reading frame 84
There are no transcripts associated with this gene.

Gene
CCDC36
Gene Name
coiled-coil domain containing 36
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135197.1 9 Intron NP_001128669.1
NM_178173.3 9 UTR 5 NP_835467.2
XM_006713124.3 9 Intron XP_006713187.1
XM_011533670.2 9 Intron XP_011531972.1
XM_011533671.2 9 Intron XP_011531973.1
XM_011533672.2 9 Intron XP_011531974.1
XM_011533673.2 9 Intron XP_011531975.1

View Full Product Details