Product Details

SNP ID
rs112980203
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:52813471 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCACTCCCGGGGGCCCCTCCT[C/G]GTAATCCAGCCTGCGCTCTCTGAAA
Phenotype
MIM: 146650 MIM: 600564
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ITIH3 PubMed Links

Gene Details

Gene
ITIH3
Gene Name
inter-alpha-trypsin inhibitor heavy chain 3
There are no transcripts associated with this gene.

Gene
ITIH4
Gene Name
inter-alpha-trypsin inhibitor heavy chain family member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166449.1 2712 Missense Mutation CAG,GAG Q885E NP_001159921.1
NM_002218.4 2712 Missense Mutation CAG,GAG Q915E NP_002209.2

View Full Product Details