Product Details

SNP ID
rs113498903
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:55953186 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGATTTTGTTTTGGAACCCTATA[A/G]ACTTGAAAATGCAAGATTGAGTAGA
Phenotype
MIM: 611423
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CEP135 PubMed Links

Gene Details

Gene
CEP135
Gene Name
centrosomal protein 135
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025009.4 483 Missense Mutation AAA,AGA K72R NP_079285.2
XM_005265788.3 483 UTR 5 XP_005265845.1
XM_006714055.2 483 Missense Mutation AAA,AGA K72R XP_006714118.1
XM_011534412.2 483 Intron XP_011532714.1

View Full Product Details