Product Details

SNP ID
rs115847385
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:157734497 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGTGTACTCTCTGTGGTACCCA[C/T]AGGTCTTATCATGGTGTTGAACTAA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
LSM11 PubMed Links

Gene Details

Gene
LSM11
Gene Name
LSM11, U7 small nuclear RNA associated
There are no transcripts associated with this gene.

Gene
THG1L
Gene Name
tRNA-histidine guanylyltransferase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317824.1 Intron NP_001304753.1
NM_001317825.1 Intron NP_001304754.1
NM_001317826.1 Intron NP_001304755.1
NM_017872.4 Intron NP_060342.2
XM_017009603.1 Intron XP_016865092.1

View Full Product Details