Product Details

SNP ID
rs111753822
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:31739566 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAACTTACTACATCAGAACTCCT[C/G]CTGAGGTATAAGAGGATACGTCTAA
Phenotype
MIM: 606655
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
RXFP2 PubMed Links

Gene Details

Gene
RXFP2
Gene Name
relaxin/insulin like family peptide receptor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001166058.1 25 UTR 5 NP_001159530.1
NM_130806.3 25 UTR 5 NP_570718.1
XM_017020389.1 25 UTR 5 XP_016875878.1

View Full Product Details