Product Details

SNP ID
rs115704657
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.6:156782350 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTAATTGGGACAAAACGTGTATAT[A/G]TTATAGGAGAAGGGTCTTTTTAAAA
Phenotype
MIM: 614556
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ARID1B PubMed Links

Gene Details

Gene
ARID1B
Gene Name
AT-rich interaction domain 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017519.2 Intron NP_059989.2
NM_020732.3 Intron NP_065783.3
XM_005267069.4 Intron XP_005267126.2
XM_011535984.2 Intron XP_011534286.2
XM_011535988.2 Intron XP_011534290.1
XM_017011103.1 Intron XP_016866592.1
XM_017011104.1 Intron XP_016866593.1
XM_017011105.1 Intron XP_016866594.1
XM_017011106.1 Intron XP_016866595.1
XM_017011107.1 Intron XP_016866596.1
XM_017011108.1 Intron XP_016866597.1
XM_017011109.1 Intron XP_016866598.1
Gene
MIR4466
Gene Name
microRNA 4466
There are no transcripts associated with this gene.

View Full Product Details