Product Details

SNP ID
rs114117560
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:167329760 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGCCTCTGCAATCAAGTGCCCC[A/C]GACCCTGTGGTTTTATGTGTTATTT
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TTLL2 PubMed Links

Gene Details

Gene
TTLL2
Gene Name
tubulin tyrosine ligase like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_031949.4 Intron NP_114155.4
XM_006715572.3 Intron XP_006715635.1

View Full Product Details