Product Details

SNP ID
rs112414932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:96006285 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATGCTGTGCCTTTCTGTCCATTTT[A/G]ATTTCTAAGATTTTGAGCTAGTACT
Phenotype
MIM: 609457 MIM: 151570
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HAL PubMed Links

Gene Details

Gene
HAL
Gene Name
histidine ammonia-lyase
There are no transcripts associated with this gene.

Gene
LTA4H
Gene Name
leukotriene A4 hydrolase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000895.2 Intron NP_000886.1
NM_001256643.1 Intron NP_001243572.1
NM_001256644.1 Intron NP_001243573.1
XM_005268871.1 Intron XP_005268928.1
XM_011538348.1 Intron XP_011536650.1
XM_011538349.2 Intron XP_011536651.1

View Full Product Details