Product Details
- SNP ID
-
rs113777896
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:106239080 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACAAACAACCTATCCAGATCATTCC[C/T]CAGGTCATCTAGTAAACCCTTGGCT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CKAP4
PubMed Links
Gene Details
- Gene
- CKAP4
- Gene Name
- cytoskeleton associated protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006825.3 |
1837 |
Missense Mutation |
AGG,GGG |
R585G |
NP_006816.2 |
View Full Product Details