Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001077690.1 | 2594 | UTR 3 | NP_001071158.1 | ||
NM_001077691.1 | 2594 | UTR 3 | NP_001071159.1 | ||
NM_001077692.1 | 2594 | UTR 3 | NP_001071160.1 | ||
NM_024740.2 | 2594 | UTR 3 | NP_079016.2 | ||
XM_005277723.4 | 2594 | UTR 3 | XP_005277780.1 | ||
XM_005277724.4 | 2594 | UTR 3 | XP_005277781.1 | ||
XM_006718913.2 | 2594 | Intron | XP_006718976.1 | ||
XM_011542992.1 | 2594 | Intron | XP_011541294.1 | ||
XM_017018313.1 | 2594 | Intron | XP_016873802.1 | ||
XM_017018314.1 | 2594 | UTR 3 | XP_016873803.1 | ||
XM_017018315.1 | 2594 | UTR 3 | XP_016873804.1 | ||
XM_017018316.1 | 2594 | UTR 3 | XP_016873805.1 | ||
XM_017018317.1 | 2594 | UTR 3 | XP_016873806.1 | ||
XM_017018318.1 | 2594 | UTR 3 | XP_016873807.1 | ||
XM_017018319.1 | 2594 | UTR 3 | XP_016873808.1 | ||
XM_017018320.1 | 2594 | UTR 3 | XP_016873809.1 | ||
XM_017018321.1 | 2594 | UTR 3 | XP_016873810.1 | ||
XM_017018322.1 | 2594 | UTR 3 | XP_016873811.1 | ||
XM_017018323.1 | 2594 | Intron | XP_016873812.1 | ||
XM_017018324.1 | 2594 | Intron | XP_016873813.1 | ||
XM_017018325.1 | 2594 | UTR 3 | XP_016873814.1 | ||
XM_017018326.1 | 2594 | UTR 3 | XP_016873815.1 |