Product Details

SNP ID
rs113986817
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:106010535 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCACCTGCATCCCTGAAATCTGCC[A/T]CATTTTGCCAGTCAAAATTTGCATC
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
MSANTD4 PubMed Links

Gene Details

Gene
MSANTD4
Gene Name
Myb/SANT DNA binding domain containing 4 with coiled-coils
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318747.1 985 Missense Mutation GAG,GTG E128V NP_001305676.1
NM_001318748.1 985 Missense Mutation GAG,GTG E128V NP_001305677.1
NM_001318749.1 985 Missense Mutation GAG,GTG E128V NP_001305678.1
NM_001318750.1 985 Missense Mutation GAG,GTG E128V NP_001305679.1
NM_032424.2 985 Missense Mutation GAG,GTG E128V NP_115800.1
XM_011543022.2 985 Missense Mutation GAG,GTG E128V XP_011541324.1
XM_017018416.1 985 Missense Mutation GAG,GTG E128V XP_016873905.1
XM_017018417.1 985 Missense Mutation GAG,GTG E128V XP_016873906.1

View Full Product Details