Product Details

SNP ID
rs117349341
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:812235 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAATGTGCTTTGAATTTATATTACA[A/T]AATAGCCACTTGAAACTGGAGTGAG
Phenotype
MIM: 616513
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
LARP4B PubMed Links

Gene Details

Gene
LARP4B
Gene Name
La ribonucleoprotein domain family member 4B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015155.2 3225 UTR 3 NP_055970.1
XM_005252432.3 3225 Intron XP_005252489.1
XM_005252435.3 3225 Intron XP_005252492.1
XM_011519435.2 3225 Intron XP_011517737.1
XM_017015987.1 3225 UTR 3 XP_016871476.1
XM_017015988.1 3225 UTR 3 XP_016871477.1
XM_017015989.1 3225 UTR 3 XP_016871478.1
XM_017015990.1 3225 Intron XP_016871479.1
XM_017015991.1 3225 Intron XP_016871480.1
XM_017015992.1 3225 UTR 3 XP_016871481.1
XM_017015993.1 3225 UTR 3 XP_016871482.1
XM_017015994.1 3225 UTR 3 XP_016871483.1
XM_017015995.1 3225 Intron XP_016871484.1

View Full Product Details