Product Details

SNP ID
rs140120750
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:990257 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AACAGATGACTTATCTAAAAACTGG[A/G]TGATGGTAATTGTTTGGTGATTGGA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GTPBP4 PubMed Links

Gene Details

Gene
GTPBP4
Gene Name
GTP binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012341.2 Intron NP_036473.2

View Full Product Details