Product Details

SNP ID
rs140185624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:84222079 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCTTTCCTGCGATGAAGACAGCC[A/G]CGCAGGACCCACTTGCTGCAGCTGC
Phenotype
MIM: 609502 MIM: 616103
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CDHR1 PubMed Links

Gene Details

Gene
CDHR1
Gene Name
cadherin related family member 1
There are no transcripts associated with this gene.

Gene
LRIT1
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 1
There are no transcripts associated with this gene.

Gene
LRIT2
Gene Name
leucine rich repeat, Ig-like and transmembrane domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017924.3 1563 Silent Mutation CGC,CGT R498R NP_001017924.1
NM_001284223.1 1563 Silent Mutation CGC,CGT R508R NP_001271152.1

View Full Product Details