Product Details

SNP ID
rs140865131
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:133267373 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACCTGCTTTGGTGCCTGCCGGGT[A/G]TAGACAGGAACTGGGAAGGGTGGGC
Phenotype
MIM: 602267
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAM8 PubMed Links

Gene Details

Gene
ADAM8
Gene Name
ADAM metallopeptidase domain 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001109.4 2565 Silent Mutation TAC,TAT Y766Y NP_001100.3
NM_001164489.1 2565 Missense Mutation CAC,TAC H711Y NP_001157961.1
NM_001164490.1 2565 Silent Mutation TAC,TAT Y701Y NP_001157962.1
XM_011539117.2 2565 Silent Mutation TAC,TAT Y831Y XP_011537419.2
XM_017015465.1 2565 Silent Mutation TAC,TAT Y831Y XP_016870954.1
XM_017015466.1 2565 Intron XP_016870955.1

View Full Product Details