Product Details

SNP ID
rs140955773
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:102481815 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATGGAAGCCTAGTTCCACCCAGG[C/T]CAGGCCCCACCATGCTCCTACCCTG
Phenotype
MIM: 605143
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTR1A PubMed Links

Gene Details

Gene
ACTR1A
Gene Name
ARP1 actin-related protein 1 homolog A, centractin alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005736.3 Intron NP_005727.1
Gene
MFSD13A
Gene Name
major facilitator superfamily domain containing 13A
There are no transcripts associated with this gene.

View Full Product Details