Product Details

SNP ID
rs142016854
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:92834205 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCCTAGTGACCTCAAAATCACAT[C/T]CCATAGCACATTTCATTTTTCCTGG
Phenotype
MIM: 609672
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EXOC6 PubMed Links

Gene Details

Gene
EXOC6
Gene Name
exocyst complex component 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001013848.3 Intron NP_001013870.1
NM_001319194.1 Intron NP_001306123.1
NM_001319195.1 Intron NP_001306124.1
NM_001319200.1 Intron NP_001306129.1
NM_019053.5 Intron NP_061926.3
XM_017016344.1 Intron XP_016871833.1
XM_017016345.1 Intron XP_016871834.1
XM_017016346.1 Intron XP_016871835.1
XM_017016347.1 Intron XP_016871836.1
XM_017016348.1 Intron XP_016871837.1

View Full Product Details