Product Details

SNP ID
rs142030036
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:101035558 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGGAGGGCCAAGTATGTGTAT[A/G]ACTCCGCCTTCCATCCGGACACAGG
Phenotype
MIM: 612971 MIM: 615571
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDZD7 PubMed Links

Gene Details

Gene
PDZD7
Gene Name
PDZ domain containing 7
There are no transcripts associated with this gene.

Gene
SFXN3
Gene Name
sideroflexin 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030971.3 691 Missense Mutation AAC,GAC N79D NP_112233.2

View Full Product Details